Product Details

SNP ID
rs147198180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:282045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGACCCAAGCCTGCCCAATGTG[A/C]AGGTGACCAGGCTGACACTCCTGTC
Phenotype
MIM: 602844 MIM: 603816 MIM: 608012 MIM: 603895
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGDIG PubMed Links

Gene Details

Gene
ARHGDIG
Gene Name
Rho GDP dissociation inhibitor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001176.3 349 Missense Mutation AAG,CAG K92Q NP_001167.2
Gene
AXIN1
Gene Name
axin 1
There are no transcripts associated with this gene.

Gene
PDIA2
Gene Name
protein disulfide isomerase family A member 2
There are no transcripts associated with this gene.

Gene
RGS11
Gene Name
regulator of G-protein signaling 11
There are no transcripts associated with this gene.

View Full Product Details