Product Details

SNP ID
rs147367477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30762620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGAAAAGAAGCTGAGTCGTGA[C/G]GAGAAGACCACCACGACTCTTATCG
Phenotype
MIM: 172471 MIM: 607700
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC189 PubMed Links

Gene Details

Gene
CCDC189
Gene Name
coiled-coil domain containing 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014979.2 236 Intron NP_001014979.2
NM_001195620.1 236 Intron NP_001182549.1
XM_011545976.1 236 Intron XP_011544278.1
XM_011545978.1 236 Intron XP_011544280.1
XM_017023852.1 236 Intron XP_016879341.1
XM_017023853.1 236 Intron XP_016879342.1
XM_017023854.1 236 Intron XP_016879343.1
XM_017023855.1 236 Intron XP_016879344.1
XM_017023856.1 236 Intron XP_016879345.1
XM_017023857.1 236 Intron XP_016879346.1
XM_017023858.1 236 Intron XP_016879347.1
XM_017023859.1 236 Intron XP_016879348.1
XM_017023860.1 236 Intron XP_016879349.1
XM_017023861.1 236 Intron XP_016879350.1
XM_017023862.1 236 Intron XP_016879351.1
Gene
PHKG2
Gene Name
phosphorylase kinase catalytic subunit gamma 2
There are no transcripts associated with this gene.

Gene
RNF40
Gene Name
ring finger protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207033.1 236 Missense Mutation GAC,GAG D25E NP_001193962.1
NM_001207034.1 236 Missense Mutation GAC,GAG D25E NP_001193963.1
NM_001286572.2 236 Missense Mutation GAC,GAG D25E NP_001273501.1
NM_014771.3 236 Missense Mutation GAC,GAG D25E NP_055586.1
XM_011545997.1 236 Missense Mutation GAC,GAG D25E XP_011544299.1

View Full Product Details