Product Details

SNP ID
rs148016476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30571474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGTAGGTCTCCTGCATCACGTC[C/G]CGGTACAGAGCCCTCTGCGCGGGCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF688 PubMed Links

Gene Details

Gene
ZNF688
Gene Name
zinc finger protein 688
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024683.1 934 Intron NP_001019854.1
NM_145271.3 934 Silent Mutation CGC,CGG R52R NP_660314.1
XM_005255139.2 934 Intron XP_005255196.1
XM_005255140.2 934 UTR 5 XP_005255197.1
Gene
ZNF785
Gene Name
zinc finger protein 785
There are no transcripts associated with this gene.

View Full Product Details