Product Details

SNP ID
rs148026549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66939267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAGAGTTTCTTAGCCAGTTCA[A/G]CATGACCTTCCCTTCCGACTCCATG
Phenotype
MIM: 605278 MIM: 614778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CES2 PubMed Links

Gene Details

Gene
CES2
Gene Name
carboxylesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003869.5 1524 Missense Mutation AAC,AGC N175S NP_003860.2
NM_198061.2 1524 Missense Mutation AAC,AGC N175S NP_932327.1
XM_011523421.2 1524 Missense Mutation AAC,AGC N18S XP_011521723.1
XM_017023818.1 1524 Missense Mutation AAC,AGC N18S XP_016879307.1
Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

View Full Product Details