Product Details

SNP ID
rs148094437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2855780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCAGGCAACACCCACCTTCTGG[C/G]ACCGAGAGCCAGGGTTCCCCAGCTG
Phenotype
MIM: 609343
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS22 PubMed Links

Gene Details

Gene
PRSS22
Gene Name
protease, serine 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022119.3 419 Missense Mutation TCC,TGC S118C NP_071402.1
XM_005255473.3 419 Missense Mutation TCC,TGC S118C XP_005255530.1

View Full Product Details