Product Details

SNP ID
rs148146066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23691158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCACAGCACGGAGCAGGTCTC[A/G]CACTGATGTCCCCTTATAGGACCGG
Phenotype
MIM: 604034 MIM: 602098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERN2 PubMed Links

Gene Details

Gene
ERN2
Gene Name
endoplasmic reticulum to nucleus signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308220.1 2696 Missense Mutation NP_001295149.2
NM_033266.3 2696 Missense Mutation NP_150296.4
XM_011545708.1 2696 Nonsense Mutation XP_011544010.1
XM_011545709.1 2696 Nonsense Mutation XP_011544011.1
XM_011545711.2 2696 Intron XP_011544013.1
XM_011545712.2 2696 Intron XP_011544014.1
XM_011545713.1 2696 Intron XP_011544015.1
Gene
PLK1
Gene Name
polo like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005030.5 2696 Intron NP_005021.2

View Full Product Details