Product Details

SNP ID
rs148214926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31219307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGGAGACAGTGCGTCAGTTC[C/T]GGGGGGCCGTGGGGGAGCAGCTGGG
Phenotype
MIM: 615700 MIM: 613288
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYDC1 PubMed Links

Gene Details

Gene
PYDC1
Gene Name
pyrin domain containing 1
There are no transcripts associated with this gene.

Gene
TRIM72
Gene Name
tripartite motif containing 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008274.3 789 Missense Mutation CGG,TGG R169W NP_001008275.2
XM_017023254.1 789 Missense Mutation CGG,TGG R25W XP_016878743.1

View Full Product Details