Product Details

SNP ID
rs149138180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4797465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTCCTGGGTGCTGTGATCAGA[A/G]GGGTCTGTAGCTCCAGTAGCTGGAG
Phenotype
MIM: 610660 MIM: 614574 MIM: 611562
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
There are no transcripts associated with this gene.

Gene
ROGDI
Gene Name
rogdi homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024589.2 1238 Missense Mutation CTC,TTC L287F NP_078865.1
XM_006720947.3 1238 Missense Mutation CTC,TTC L294F XP_006721010.1
XM_006720948.3 1238 Missense Mutation CTC,TTC L204F XP_006721011.1
Gene
SEPT12
Gene Name
septin 12
There are no transcripts associated with this gene.

Gene
SMIM22
Gene Name
small integral membrane protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253790.1 1238 Intron NP_001240719.1
NM_001253791.1 1238 Intron NP_001240720.1
NM_001253793.1 1238 Intron NP_001240722.1
NM_001253794.1 1238 Intron NP_001240723.1
XM_011522499.2 1238 Intron XP_011520801.1
XM_011522500.2 1238 Intron XP_011520802.1
XM_011522501.2 1238 Intron XP_011520803.1

View Full Product Details