Product Details

SNP ID
rs149157381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28932468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACTCAGCCTGGGGCTGCCAGGC[C/G]TGGGAATCCACATGAGGCCCCTGGC
Phenotype
MIM: 107265 MIM: 611869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD19 PubMed Links

Gene Details

Gene
CD19
Gene Name
CD19 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178098.1 273 Missense Mutation CTG,GTG L71V NP_001171569.1
NM_001770.5 273 Missense Mutation CTG,GTG L71V NP_001761.3
XM_006721103.3 273 Intron XP_006721166.1
XM_011545981.1 273 Missense Mutation CTG,GTG L71V XP_011544283.1
XM_017023893.1 273 Intron XP_016879382.1
Gene
RABEP2
Gene Name
rabaptin, RAB GTPase binding effector protein 2
There are no transcripts associated with this gene.

View Full Product Details