Product Details

SNP ID
rs149254133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82035487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGTCCCCACAGTACTATGTGG[G/T]ACAGTATTTTGCAAATACAAGAAGA
Phenotype
MIM: 109685
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B2 PubMed Links

Gene Details

Gene
HSD17B2
Gene Name
hydroxysteroid 17-beta dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002153.2 235 Silent Mutation GGG,GGT G21G NP_002144.1

View Full Product Details