Product Details

SNP ID
rs149261658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28499732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACAGCAGCAGCAACTCCCGCAC[A/G]GCCCGAGATAAGACGAGCTCCAAGG
Phenotype
MIM: 605220 MIM: 607042 MIM: 608273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOBR PubMed Links

Gene Details

Gene
APOBR
Gene Name
apolipoprotein B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018690.3 674 Intron NP_061160.3
Gene
CLN3
Gene Name
CLN3, battenin
There are no transcripts associated with this gene.

Gene
IL27
Gene Name
interleukin 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145659.3 674 Silent Mutation GCC,GCT A217A NP_663634.2
XM_011545780.2 674 Silent Mutation GCC,GCT A219A XP_011544082.1

View Full Product Details