Product Details

SNP ID
rs149441644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20031914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCTTGAGCGTGGCGGCTGCCA[C/T]GGTGCGGAACCGCTTGCTGATGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR139 PubMed Links

Gene Details

Gene
GPR139
Gene Name
G protein-coupled receptor 139
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002911.3 1045 Missense Mutation ATG,GTG M295V NP_001002911.1
NM_001318483.1 1045 Missense Mutation ATG,GTG M202V NP_001305412.1

View Full Product Details