Product Details

SNP ID
rs149561184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56451649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCCAGCGGAGCCCGGCCCAGCC[C/T]GGGTGGGAAAAAAGGGAAAGAAGGA
Phenotype
MIM: 604978 MIM: 615857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 170 Intron NP_008937.1
Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324357.1 170 Silent Mutation CGG,TGG R13W NP_001311286.1
NM_001324358.1 170 Intron NP_001311287.1
NM_001324359.1 170 Intron NP_001311288.1
NM_001324360.1 170 UTR 5 NP_001311289.1
NM_001324361.1 170 Intron NP_001311290.1
NM_001324362.1 170 Intron NP_001311291.1
NM_001324363.1 170 Silent Mutation CGG,TGG R13W NP_001311292.1
NM_018233.3 170 Silent Mutation CGG,TGG R13W NP_060703.3

View Full Product Details