Product Details
- SNP ID
-
rs149584051
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:84122289 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCTGGAACAAAACTCTTCCAAAG[G/T]AAACAAACAACTACTTTTATTGTTA
- Phenotype
-
MIM: 603355
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HSDL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs531441713] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSDL1
- Gene Name
- hydroxysteroid dehydrogenase like 1
- Gene
- LOC729887
- Gene Name
- uncharacterized LOC729887
There are no transcripts associated with this gene.
- Gene
- MBTPS1
- Gene Name
- membrane bound transcription factor peptidase, site 1
There are no transcripts associated with this gene.
View Full Product Details