Product Details

SNP ID
rs149613346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:685938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCTCCGTGCATCTCCTTTGCTGC[C/G]GTCCAGCCGCGTCTCACTGCCCAAC
Phenotype
MIM: 609082 MIM: 603264 MIM: 607207
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL16 PubMed Links

Gene Details

Gene
FBXL16
Gene Name
F-box and leucine rich repeat protein 16
There are no transcripts associated with this gene.

Gene
JMJD8
Gene Name
jumonji domain containing 8
There are no transcripts associated with this gene.

Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
There are no transcripts associated with this gene.

Gene
STUB1
Gene Name
STIP1 homology and U-box containing protein 1
There are no transcripts associated with this gene.

Gene
WDR24
Gene Name
WD repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032259.3 2792 Missense Mutation CGC,GGC R502G NP_115635.1
XM_011522699.2 2792 Missense Mutation CGC,GGC R591G XP_011521001.1

View Full Product Details