Product Details

SNP ID
rs149935646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2151726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACAGCTGGTCAGGAGCGGTTCC[A/G]CAGTGTTACCCATGCCTACTACCGG
Phenotype
MIM: 605455 MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB26 PubMed Links

Gene Details

Gene
RAB26
Gene Name
RAB26, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308053.1 553 Missense Mutation CAC,CGC H61R NP_001294982.1
NM_014353.4 553 Missense Mutation CAC,CGC H127R NP_055168.2
XM_011522448.1 553 Missense Mutation CAC,CGC H127R XP_011520750.1
XM_011522450.2 553 Missense Mutation CAC,CGC H11R XP_011520752.1
Gene
SNHG19
Gene Name
small nucleolar RNA host gene 19
There are no transcripts associated with this gene.

Gene
SNORD60
Gene Name
small nucleolar RNA, C/D box 60
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
There are no transcripts associated with this gene.

View Full Product Details