Product Details

SNP ID
rs149984715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:17105648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCTAAAGAATGCTTCCTTCTC[A/C]ACAAACCAATACCCCAAGGCCCACC
Phenotype
MIM: 608124
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
XYLT1 PubMed Links

Gene Details

Gene
XYLT1
Gene Name
xylosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022166.3 6012 UTR 3 NP_071449.1
XM_017023539.1 6012 Intron XP_016879028.1
XM_017023540.1 6012 Intron XP_016879029.1

View Full Product Details