Product Details

SNP ID
rs150193781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85903141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTTCCGGATCCCTTGGAAACA[C/T]GCTGGCAAGCAAGATTATAATCAGG
Phenotype
MIM: 601565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRF8 PubMed Links

Gene Details

Gene
IRF8
Gene Name
interferon regulatory factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002163.2 183 Silent Mutation CAC,CAT H42H NP_002154.1
XM_017023199.1 183 UTR 5 XP_016878688.1

View Full Product Details