Product Details

SNP ID
rs150385852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4511102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGAGACCAACGTGTGCCCTGGC[C/T]CAGTCACAGCTGGAAATCCCAGGGC
Phenotype
MIM: 610503 MIM: 141251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDIP1 PubMed Links

Gene Details

Gene
CDIP1
Gene Name
cell death-inducing p53 target 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199054.1 2078 UTR 3 NP_001185983.1
NM_001199055.1 2078 UTR 3 NP_001185984.1
NM_001199056.1 2078 UTR 3 NP_001185985.1
NM_013399.2 2078 UTR 3 NP_037531.2
Gene
HMOX2
Gene Name
heme oxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127204.1 2078 Intron NP_001120676.1
NM_001127205.1 2078 Intron NP_001120677.1
NM_001127206.2 2078 Intron NP_001120678.1
NM_001286267.1 2078 Intron NP_001273196.1
NM_001286268.1 2078 Intron NP_001273197.1
NM_001286269.1 2078 Intron NP_001273198.1
NM_001286270.1 2078 Intron NP_001273199.1
NM_001286271.1 2078 Intron NP_001273200.1
NM_002134.3 2078 Intron NP_002125.3
XM_011522473.1 2078 Intron XP_011520775.1
XM_017023196.1 2078 Intron XP_016878685.1
XM_017023197.1 2078 Intron XP_016878686.1

View Full Product Details