Product Details

SNP ID
rs150386538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2793874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTTTTCTTTCAGGAATTTTATT[A/C]ACACACTGGGAATTTCTAGGAATTT
Phenotype
MIM: 613797
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRSS33 PubMed Links

Gene Details

Gene
PRSS33
Gene Name
protease, serine 33
There are no transcripts associated with this gene.

Gene
PRSS41
Gene Name
protease, serine 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135086.1 Intron NP_001128558.1
XM_011522477.2 Intron XP_011520779.1

View Full Product Details