Product Details

SNP ID
rs150748596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:51137421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATCCCCACTTCCTGATCTTGC[C/T]GCCAAATCCTTCTGGAACATTTCTG
Phenotype
MIM: 602218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371253 PubMed Links

Gene Details

Gene
LOC105371253
Gene Name
uncharacterized LOC105371253
There are no transcripts associated with this gene.

Gene
SALL1
Gene Name
spalt like transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127892.1 3697 Silent Mutation GCA,GCG A1125A NP_001121364.1
NM_002968.2 3697 Silent Mutation GCA,GCG A1222A NP_002959.2
XM_006721241.3 3697 Silent Mutation GCA,GCG A1222A XP_006721304.1
XM_011523254.2 3697 Silent Mutation GCA,GCG A1222A XP_011521556.1

View Full Product Details