Product Details

SNP ID
rs150884036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14648440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATTAGACTGAGATTTGAAGAC[A/G]TTCAGCAGAATAATGACATAGTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BFAR PubMed Links

Gene Details

Gene
BFAR
Gene Name
bifunctional apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016561.2 597 Missense Mutation ATT,GTT I106V NP_057645.1
XM_005255350.2 597 Intron XP_005255407.1
XM_011522520.1 597 Intron XP_011520822.1

View Full Product Details