Product Details

SNP ID
rs150887497
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21962817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACAGCACCAGAATTTCGTCGTT[C/G]GGAAGTAGCTGACCTTCAGCCTCAG
Phenotype
MIM: 191329
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PDZD9 PubMed Links
Additional Information
For this assay, SNP(s) [rs2228473] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PDZD9
Gene Name
PDZ domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173806.3 806 Intron NP_776167.2
XM_006721029.3 806 Intron XP_006721092.1
XM_011545785.2 806 Intron XP_011544087.1
XM_017023109.1 806 Intron XP_016878598.1
XM_017023110.1 806 Intron XP_016878599.1
Gene
UQCRC2
Gene Name
ubiquinol-cytochrome c reductase core protein II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003366.3 806 Missense Mutation TCG,TGG S149W NP_003357.2

View Full Product Details