Product Details

SNP ID
rs150893487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23065678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGATCCCATGGAAAACCAAAG[A/G]GATTTGTAGAAACGTTTTTCCTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP31 PubMed Links

Gene Details

Gene
USP31
Gene Name
ubiquitin specific peptidase 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020718.3 5544 UTR 3 NP_065769.3
XM_017023496.1 5544 UTR 3 XP_016878985.1
XM_017023497.1 5544 UTR 3 XP_016878986.1
XM_017023498.1 5544 UTR 3 XP_016878987.1
XM_017023499.1 5544 UTR 3 XP_016878988.1
XM_017023500.1 5544 Intron XP_016878989.1

View Full Product Details