Product Details

SNP ID
rs150963331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2152847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGACCGAGATCCACGAGTAC[A/G]CCCAGCACGACGTGGCGCTCATGCT
Phenotype
MIM: 605455 MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB26 PubMed Links

Gene Details

Gene
RAB26
Gene Name
RAB26, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308053.1 669 Missense Mutation ACC,GCC T100A NP_001294982.1
NM_014353.4 669 Missense Mutation ACC,GCC T166A NP_055168.2
XM_011522448.1 669 Missense Mutation ACC,GCC T166A XP_011520750.1
XM_011522450.2 669 Missense Mutation ACC,GCC T50A XP_011520752.1
Gene
SNHG19
Gene Name
small nucleolar RNA host gene 19
There are no transcripts associated with this gene.

Gene
SNORD60
Gene Name
small nucleolar RNA, C/D box 60
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
There are no transcripts associated with this gene.

View Full Product Details