Product Details

SNP ID
rs151044832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10591989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAGTAAGGGGTGGAGCACTAGG[C/T]GACTGTATGTCCAGGGGAGTTGGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEKT5 PubMed Links

Gene Details

Gene
TEKT5
Gene Name
tektin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144674.1 Intron NP_653275.1
XM_011522383.2 Intron XP_011520685.1
XM_011522385.2 Intron XP_011520687.1
XM_011522389.1 Intron XP_011520691.1
XM_011522390.2 Intron XP_011520692.1
XM_017022955.1 Intron XP_016878444.1
XM_017022956.1 Intron XP_016878445.1
XM_017022957.1 Intron XP_016878446.1
XM_017022958.1 Intron XP_016878447.1

View Full Product Details