Product Details

SNP ID
rs151136336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4260204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGAGGAGGCGGTGCTGGCAC[A/G]ATCACCGTGGGGTGGTGGGTGGGGG
Phenotype
MIM: 600743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01569 PubMed Links

Gene Details

Gene
LINC01569
Gene Name
long intergenic non-protein coding RNA 1569
There are no transcripts associated with this gene.

Gene
LOC105371063
Gene Name
putative uncharacterized protein encoded by LINC00596
There are no transcripts associated with this gene.

Gene
TFAP4
Gene Name
transcription factor AP-4 (activating enhancer binding protein 4)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003223.2 1098 Silent Mutation ATC,ATT I236I NP_003214.1
XM_011522633.2 1098 Silent Mutation ATC,ATT I223I XP_011520935.1
XM_011522635.2 1098 Silent Mutation ATC,ATT I176I XP_011520937.1

View Full Product Details