Product Details

SNP ID
rs3815561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41040924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCGGCAACAGGACTGTCCACA[G/A]TAGGATGGGCGGCAGCAGGAGGCCT
Phenotype
MIM: 608819 MIM: 608820
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KRTAP1-1 PubMed Links

Gene Details

Gene
KRTAP1-1
Gene Name
keratin associated protein 1-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030967.2 538 Silent Mutation TAC,TAT Y158Y NP_112229.1
Gene
KRTAP1-3
Gene Name
keratin associated protein 1-3
There are no transcripts associated with this gene.

Gene
KRTAP2-1
Gene Name
keratin associated protein 2-1
There are no transcripts associated with this gene.

View Full Product Details