Product Details

SNP ID
rs138125040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48026800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCCCACAGTGCCACGTCCAGGC[A/G]GAGACGGGGCCCAGGCCTGACACTC
Phenotype
MIM: 615526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPZ2 PubMed Links

Gene Details

Gene
COPZ2
Gene Name
coatomer protein complex subunit zeta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016429.3 Intron NP_057513.1
XM_011524874.2 Intron XP_011523176.1

View Full Product Details