Product Details

SNP ID
rs138141702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58543125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTTGTGGGAGCCCTCAGACAC[A/T]GATGGAACTGTCACATGGTGAGTTG
Phenotype
MIM: 603696
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C17orf47 PubMed Links

Gene Details

Gene
C17orf47
Gene Name
chromosome 17 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038704.2 1248 Silent Mutation TCA,TCT S354S NP_001033793.2
XM_011524637.2 1248 Silent Mutation TCA,TCT S354S XP_011522939.1
XM_017024498.1 1248 Silent Mutation TCA,TCT S354S XP_016879987.1
Gene
SEPT4
Gene Name
septin 4
There are no transcripts associated with this gene.

Gene
SEPT4-AS1
Gene Name
SEPT4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details