Product Details

SNP ID
rs138152713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50836108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAATCCCAACCTCTGGGTGGA[C/T]GCACAGAGCACCTGCAGGCGGGAGT
Phenotype
MIM: 610895
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WFIKKN2 PubMed Links

Gene Details

Gene
WFIKKN2
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175575.5 865 Silent Mutation GAC,GAT D57D NP_783165.1
XM_017024172.1 865 Intron XP_016879661.1

View Full Product Details