Product Details

SNP ID
rs138184965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36017832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACTGGACTCACCTTTTGTGAC[C/G]CGGGCCTGGGATCCAAGGGCAGTAA
Phenotype
MIM: 602494
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCL23 PubMed Links

Gene Details

Gene
CCL23
Gene Name
C-C motif chemokine ligand 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005064.5 137 Silent Mutation CGC,CGG R22R NP_005055.3
NM_145898.3 137 Silent Mutation CGC,CGG R22R NP_665905.2

View Full Product Details