Product Details

SNP ID
rs138266618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74432110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGAAACTCCCATCTCCCTCACC[A/C]GCCGGAAAGTACGAGTCGGCTCAGC
Phenotype
MIM: 605949
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPRC5C PubMed Links

Gene Details

Gene
GPRC5C
Gene Name
G protein-coupled receptor class C group 5 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018653.3 583 Intron NP_061123.3
NM_022036.2 583 Silent Mutation CCA,CCC P24P NP_071319.2
XM_005257512.1 583 Intron XP_005257569.1
XM_005257514.1 583 Silent Mutation CCA,CCC P24P XP_005257571.1
XM_017024855.1 583 Intron XP_016880344.1
XM_017024856.1 583 Intron XP_016880345.1

View Full Product Details