Product Details
- SNP ID
-
rs138306277
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7890649 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGGGGGCAGTGAATATGGAACC[A/G]GACCGGGTCGGAAACGAAGAAGGAA
- Phenotype
-
MIM: 602120
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHD3
PubMed Links
Gene Details
- Gene
- CHD3
- Gene Name
- chromodomain helicase DNA binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005271.2 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
NP_001005271.2 |
NM_001005273.2 |
512 |
Missense Mutation |
AGA,GGA |
R98G |
NP_001005273.1 |
NM_005852.3 |
512 |
Missense Mutation |
AGA,GGA |
R98G |
NP_005843.2 |
XM_005256427.4 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_005256484.1 |
XM_005256428.4 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_005256485.1 |
XM_005256429.4 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_005256486.1 |
XM_005256431.4 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_005256488.1 |
XM_006721423.3 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_006721486.1 |
XM_006721424.3 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_006721487.1 |
XM_006721428.3 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_006721491.1 |
XM_017024061.1 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_016879550.1 |
XM_017024062.1 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_016879551.1 |
XM_017024063.1 |
512 |
Missense Mutation |
AGA,GGA |
R98G |
XP_016879552.1 |
XM_017024064.1 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_016879553.1 |
XM_017024065.1 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_016879554.1 |
XM_017024066.1 |
512 |
Missense Mutation |
AGA,GGA |
R78G |
XP_016879555.1 |
XM_017024067.1 |
512 |
Missense Mutation |
AGA,GGA |
R87G |
XP_016879556.1 |
XM_017024068.1 |
512 |
Missense Mutation |
AGA,GGA |
R78G |
XP_016879557.1 |
XM_017024069.1 |
512 |
UTR 5 |
|
|
XP_016879558.1 |
XM_017024070.1 |
512 |
Missense Mutation |
AGA,GGA |
R157G |
XP_016879559.1 |
- Gene
- NAA38
- Gene Name
- N(alpha)-acetyltransferase 38, NatC auxiliary subunit
There are no transcripts associated with this gene.
View Full Product Details