Product Details

SNP ID
rs138336061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74210576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGGGGAGGTGCATGGGTTGGGG[C/T]GGCCACCAGGTGGTGCTGTGCCACA
Phenotype
MIM: 604182 MIM: 608855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGC16275 PubMed Links

Gene Details

Gene
MGC16275
Gene Name
uncharacterized protein MGC16275
There are no transcripts associated with this gene.

Gene
RPL38
Gene Name
ribosomal protein L38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000999.3 Intron NP_000990.1
NM_001035258.1 Intron NP_001030335.1
Gene
TTYH2
Gene Name
tweety family member 2
There are no transcripts associated with this gene.

View Full Product Details