Product Details

SNP ID
rs138376837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32021428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCAGGAACCAACTGAAAATTT[A/G]GCTCCATTCCTGAAGGAATTGGATT
Phenotype
MIM: 616558
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC37B PubMed Links

Gene Details

Gene
LRRC37B
Gene Name
leucine rich repeat containing 37B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321350.1 487 Silent Mutation TTA,TTG L12L NP_001308279.1
NM_052888.2 487 Intron NP_443120.2

View Full Product Details