Product Details

SNP ID
rs138392672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82322249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGTTTGAACACCAGTGCCAGC[A/G]GCCTTGGGGTGGGCTCGGAGTCTGG
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
There are no transcripts associated with this gene.

Gene
SECTM1
Gene Name
secreted and transmembrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003004.2 1088 Missense Mutation CCG,CTG P220L NP_002995.1
XM_005256392.3 1088 Missense Mutation CCG,CTG P220L XP_005256449.1
XM_011523588.2 1088 Missense Mutation CCG,CTG P220L XP_011521890.1

View Full Product Details