Product Details
- SNP ID
-
rs138760396
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:39463037 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGAAGTGGCTCTTACAGCGGGCG[A/G]TCGCCCAGTCCCTATGGTCGAAGGC
- Phenotype
-
MIM: 615514
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDK12
PubMed Links
Gene Details
- Gene
- CDK12
- Gene Name
- cyclin dependent kinase 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015083.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
NP_055898.1 |
NM_016507.3 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
NP_057591.2 |
XM_005257456.3 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_005257513.1 |
XM_005257458.4 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_005257515.1 |
XM_011524892.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523194.1 |
XM_011524893.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523195.1 |
XM_011524894.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523196.1 |
XM_011524895.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523197.1 |
XM_011524896.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523198.1 |
XM_011524897.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523199.1 |
XM_011524898.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523200.1 |
XM_011524899.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523201.1 |
XM_011524900.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523202.1 |
XM_011524901.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523203.1 |
XM_011524902.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523204.1 |
XM_011524903.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523205.1 |
XM_011524905.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523207.1 |
XM_011524906.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523208.1 |
XM_011524907.2 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_011523209.1 |
XM_017024744.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880233.1 |
XM_017024745.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880234.1 |
XM_017024746.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880235.1 |
XM_017024747.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880236.1 |
XM_017024748.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880237.1 |
XM_017024749.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880238.1 |
XM_017024750.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880239.1 |
XM_017024751.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880240.1 |
XM_017024752.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880241.1 |
XM_017024753.1 |
1527 |
Silent Mutation |
CGA,CGG |
R322R |
XP_016880242.1 |
View Full Product Details