Product Details

SNP ID
rs138772502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28578086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCGAAGGTTCTCCTCTAGGATA[G/T]TTCTCTAGAAAGCAAACAGATTTTA
Phenotype
MIM: 103870 MIM: 610271 MIM: 615562
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDOC PubMed Links

Gene Details

Gene
ALDOC
Gene Name
aldolase, fructose-bisphosphate C
There are no transcripts associated with this gene.

Gene
PIGS
Gene Name
phosphatidylinositol glycan anchor biosynthesis class S
There are no transcripts associated with this gene.

Gene
SPAG5
Gene Name
sperm associated antigen 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006461.3 3526 Missense Mutation AAT,ACT N1145T NP_006452.3

View Full Product Details