Product Details

SNP ID
rs138806111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3440294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTCTGGTCATAGTTGCCAAC[A/G]CATCTATGAACTCGGCCTCTAATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1E2 PubMed Links

Gene Details

Gene
OR1E2
Gene Name
olfactory receptor family 1 subfamily E member 2
There are no transcripts associated with this gene.

Gene
SPATA22
Gene Name
spermatogenesis associated 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170695.1 1183 Silent Mutation TGC,TGT C315C NP_001164166.1
NM_001170696.1 1183 Silent Mutation TGC,TGT C272C NP_001164167.1
NM_001170697.1 1183 Silent Mutation TGC,TGT C315C NP_001164168.1
NM_001170698.1 1183 Silent Mutation TGC,TGT C315C NP_001164169.1
NM_001170699.1 1183 UTR 3 NP_001164170.1
NM_001321336.1 1183 Silent Mutation TGC,TGT C299C NP_001308265.1
NM_001321337.1 1183 Silent Mutation TGC,TGT C315C NP_001308266.1
NM_032598.4 1183 Silent Mutation TGC,TGT C315C NP_115987.2

View Full Product Details