Product Details

SNP ID
rs139097766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79834793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGCGAGCCTCCCCCTCTGCCAC[C/G]TCGCCGGACTTGATCTTCACCGCCT
Phenotype
MIM: 603079
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CBX4 PubMed Links

Gene Details

Gene
CBX4
Gene Name
chromobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003655.2 2521 Silent Mutation GAC,GAG D283E NP_003646.2
XM_011525399.2 2521 Missense Mutation GAC,GAG D217E XP_011523701.1

View Full Product Details