Product Details

SNP ID
rs139125476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61401712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTCCCCCCCTTCAAGGTGCGA[C/G]TCAGCGGCCTGGACAAGAAGGCCAA
Phenotype
MIM: 607470 MIM: 600747
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCAS3 PubMed Links

Gene Details

Gene
BCAS3
Gene Name
BCAS3, microtubule associated cell migration factor
There are no transcripts associated with this gene.

Gene
TBX2
Gene Name
T-box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005994.3 705 Missense Mutation CTC,GTC L142V NP_005985.3
XM_011525159.1 705 Intron XP_011523461.1
Gene
TBX2-AS1
Gene Name
TBX2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details