Product Details

SNP ID
rs139199066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37621272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTGAAACTGTGGACTGTGTT[A/C]TCTCTCTGGTTAACAGAATATATAT
Phenotype
MIM: 612500 MIM: 607291
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDX52 PubMed Links

Gene Details

Gene
DDX52
Gene Name
DEAD-box helicase 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291476.1 1400 Silent Mutation NP_001278405.1
NM_007010.4 1400 Silent Mutation NP_008941.3
XM_011524232.2 1400 Missense Mutation XP_011522534.1
XM_011524233.2 1400 Missense Mutation XP_011522535.1
Gene
MIR378J
Gene Name
microRNA 378j
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
There are no transcripts associated with this gene.

View Full Product Details