Product Details

SNP ID
rs139372330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28380471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTGCTAGCGCCAAGGCCCTAT[C/T]ACTACGATCTGGTTCACTCATTGTT
Phenotype
MIM: 607732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SARM1 PubMed Links

Gene Details

Gene
SARM1
Gene Name
sterile alpha and TIR motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015077.3 Intron NP_055892.2

View Full Product Details