Product Details

SNP ID
rs139402425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81890758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTTCCCACCTGTCTCCACGCC[C/G]GCACCACCGCCGAAGCCACTGTCGA
Phenotype
MIM: 604171 MIM: 614534
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005782.3 327 Silent Mutation GCC,GCG A107A NP_005773.3
Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002244.2 327 Intron NP_001002244.1
NM_001002245.2 327 Intron NP_001002245.1
NM_001002246.2 327 Intron NP_001002246.1
NM_001002247.2 327 Intron NP_001002247.1
NM_001002248.2 327 Intron NP_001002248.1
NM_001002249.2 327 Intron NP_001002249.1
NM_001289414.1 327 Intron NP_001276343.1
NM_001289415.1 327 Intron NP_001276344.1
NM_001289416.1 327 Intron NP_001276345.1
NM_001289417.1 327 Intron NP_001276346.1
NM_001289418.1 327 Intron NP_001276347.1
NM_001289419.1 327 Intron NP_001276348.1
NM_001289420.1 327 Intron NP_001276349.1
NM_016476.11 327 Intron NP_057560.8

View Full Product Details