Product Details

SNP ID
rs139420851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30904317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGGAAAGCTTCAAGTTCTTTAG[C/T]AGATGCTGTATTCAACACATGCAAG
Phenotype
MIM: 609534 MIM: 616422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATAD5 PubMed Links

Gene Details

Gene
ATAD5
Gene Name
ATPase family, AAA domain containing 5
There are no transcripts associated with this gene.

Gene
TEFM
Gene Name
transcription elongation factor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024683.3 798 Missense Mutation ACT,GCT T82A NP_078959.3
XM_006722084.1 798 UTR 5 XP_006722147.1

View Full Product Details