Product Details

SNP ID
rs139450838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6447695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCACATGCCCACCCATTACGGC[A/G]ATCAAGGCGGGAGGCTGCCTTCCGG
Phenotype
MIM: 608921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM64A PubMed Links

Gene Details

Gene
FAM64A
Gene Name
family with sequence similarity 64 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195228.1 1010 Intron NP_001182157.1
NM_019013.2 1010 Intron NP_061886.2
XM_011523944.2 1010 Missense Mutation CAA,CGA Q176R XP_011522246.1
XM_017024778.1 1010 Missense Mutation CAA,CGA Q130R XP_016880267.1
Gene
PITPNM3
Gene Name
PITPNM family member 3
There are no transcripts associated with this gene.

View Full Product Details