Product Details

SNP ID
rs139506094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1569623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAACACACACGCACACAGTCAG[A/C]GGCACGGTCCCGCAGCTGTATGAGG
Phenotype
MIM: 600174 MIM: 610791
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PITPNA PubMed Links

Gene Details

Gene
PITPNA
Gene Name
phosphatidylinositol transfer protein alpha
There are no transcripts associated with this gene.

Gene
SLC43A2
Gene Name
solute carrier family 43 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284498.1 7830 UTR 3 NP_001271427.1
NM_001284499.1 7830 UTR 3 NP_001271428.1
NM_001321364.1 7830 UTR 3 NP_001308293.1
NM_001321365.1 7830 UTR 3 NP_001308294.1
NM_152346.2 7830 UTR 3 NP_689559.1
XM_017024177.1 7830 Intron XP_016879666.1
XM_017024178.1 7830 Intron XP_016879667.1
XM_017024179.1 7830 Intron XP_016879668.1

View Full Product Details