Product Details
- SNP ID
-
rs139515389
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48613403 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGTCCCAGGGCGTGCGATACCTC[G/T]ATTCGCCGCTTACGAGTCAGATAGG
- Phenotype
-
MIM: 142961
MIM: 142962
MIM: 142963
MIM: 142964
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HOXB-AS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs45441492] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HOXB-AS3
- Gene Name
- HOXB cluster antisense RNA 3
There are no transcripts associated with this gene.
- Gene
- HOXB6
- Gene Name
- homeobox B6
There are no transcripts associated with this gene.
- Gene
- HOXB7
- Gene Name
- homeobox B7
There are no transcripts associated with this gene.
- Gene
- HOXB8
- Gene Name
- homeobox B8
- Gene
- HOXB9
- Gene Name
- homeobox B9
There are no transcripts associated with this gene.
View Full Product Details