Product Details

SNP ID
rs139605788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5434945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGTTGGTGGGATGCTGTTGTT[A/G]ATGTTGAAAGTGACCGTGATTCTAA
Phenotype
MIM: 601269 MIM: 614405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QBP PubMed Links

Gene Details

Gene
C1QBP
Gene Name
complement C1q binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001212.3 Intron NP_001203.1
Gene
DHX33
Gene Name
DEAH-box helicase 33
There are no transcripts associated with this gene.

Gene
RPAIN
Gene Name
RPA interacting protein
There are no transcripts associated with this gene.

View Full Product Details